Learn More
Ampco Phillips Screwdriver
SDP
Catalog No. 19174257 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
6 in.
Catalog No. Length (English)
19-174-257 6 in.
1 options
Catalog No. 19-174-257 Supplier Ampco Supplier No. S1098
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant Phillips screwdriver with cellulose acetate handle.
  • Type 0.
  • Comes with lifetime warranty.
  • Tip length: 2 and 3/8 in.
  • Total length: 6 in.
Specifications
Product Type Phillips Screwdriver
Length (English) 6 in.