Learn More
Bulbman Inc Halogen 5761 30W 6V
SDP
Catalog No. NC9057465
Encompass_Preferred
Bulb; Focusline Single Ended; 6V; 30W; 765 Lumens; G4 Base; T3.5 Shape; 31mm; 100 Life hr.; Kelvin 3200; Clear; C-6F Filament
Catalog No. NC9057465 Supplier Bulbman Inc Supplier No. 121442
Bulb; Focusline Single Ended; 6V; 30W; 765 Lumens; G4 Base; T3.5 Shape; 31mm; 100 Life hr.; Kelvin 3200; Clear; C-6F Filament