Learn More
TARGETMOL CHEMICALS INC PHILLYRIN 25MG
SDP
Catalog No. 502577038
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phillyrin (Forsythin) is a novel AMPK activator has anti-obesity effects in nutritive obesity mice it can prevent lipid accumulation in HepG2 cells by blocking the expression of SREBP-1c and FAS through LKB1/AMPK activation. Phillyrin(Forsythin) may be a new preventive agent of acute lung injury in the clinical setting it potentially contributes to the suppression of the activation of MAPK and NF-(kappa)B pathways it also has protective effects on H2O2-induced oxidative stress and apoptosis in PC12 cells. purity: 99%
Catalog No. 50-257-7038 Supplier TARGETMOL CHEMICALS INC Supplier No. T285425MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phillyrin (Forsythin) is a novel AMPK activator has anti-obesity effects in nutritive obesity mice it can prevent lipid accumulation in HepG2 cells by blocking the expression of SREBP-1c and FAS through LKB1/AMPK activation. Phillyrin(Forsythin) may be a new preventive agent of acute lung injury in the clinical setting it potentially contributes to the suppression of the activation of MAPK and NF-(kappa)B pathways it also has protective effects on H2O2-induced oxidative stress and apoptosis in PC12 cells. purity: 99%