Learn More
Medchemexpress LLC HY-N0482 5mg Medchemexpress, Phillyrin CAS:487-41-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04825MG
Encompass_Preferred
Medchemexpress, HY-N0482 5mg Phillyrin CAS:487-41-2 Phillyrin is isolated from Forsythia suspensa Vahl (Oleaceae), has antibacterial and anti-inflammatory activities. Phillyrin has potential inductive effects on rat CYP1A2 and CYP2D1 activities, without affecting CYP2C11 and CYP3A1/2 activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0276