Learn More
Cayman Chemical PhIlnthotoxIn 74hydrochLR 10mg
SDP
Catalog No. 502969978
Encompass_Preferred
A subtype-selective AMPA receptor antagonist that inhibits homomeric GluR1 and GluR3 as well as heteromeric GluR1/2 receptors (80-100% of glutamate-evoked currents inhibited at 100 µM); inhibits the GluR5(Q) receptor with a Ki value of 0.29 µM
Catalog No. 50-296-9978 Supplier Cayman Chemical Supplier No. 1460710 MG
A subtype-selective AMPA receptor antagonist that inhibits homomeric GluR1 and GluR3 as well as heteromeric GluR1/2 receptors (80-100% of glutamate-evoked currents inhibited at 100 µM); inhibits the GluR5(Q) receptor with a Ki value of 0.29 µM