Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerve Conduction Chamber
SCI_ED

Catalog No. S43927J Shop All Phipps & Bird Products
Change view
Click to view available options
Clear plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S43927J Clear plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43927J Supplier Phipps & Bird Supplier No. 7092900/EMD

missing translation for 'validMainframeMsg'

Used for studying conduction of bioelectric impulses in nerve tissue.

  • Clear Plastic chamber
  • Contains nine platinum wire electrodes with binding post for stimulating and recording

Specifications

For Use With (Application) For studying conduction of bioelectric impulses in nerve tissue
Includes Nine Platinum wire electrodes
Material Plastic
Product Type Nerve Conduction Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.