Learn More
Meso Scale Discovery pHistone HSer10 WCLPl
SDP
Catalog No. 5000081701
Encompass
The Mesoscale Discovery P-Histone H3 (Ser10) Whole Cell Lysate Kit (K150EWD) is a multiplex immunoassay for quantifying phosphorylated Histone H3 (Ser10) in cell lysates It includes a 96-well plate with capture and detection antibodies buffers calibrator and controls Using electrochemiluminescence (ECL) technology it offers high sensitivity and a broad dynamic range Suitable for studying cell proliferation mitosis apoptosis and cell cycle regulation the kit detects 50 - 50 000 pg/mL P-Histone H3 (Ser10) with a 25 L sample per well
Catalog No. 50-000-81701 Supplier Meso Scale Discovery Supplier No. K150EWD1
The Mesoscale Discovery P-Histone H3 (Ser10) Whole Cell Lysate Kit (K150EWD) is a multiplex immunoassay for quantifying phosphorylated Histone H3 (Ser10) in cell lysates It includes a 96-well plate with capture and detection antibodies buffers calibrator and controls Using electrochemiluminescence (ECL) technology it offers high sensitivity and a broad dynamic range Suitable for studying cell proliferation mitosis apoptosis and cell cycle regulation the kit detects 50 - 50 000 pg/mL P-Histone H3 (Ser10) with a 25 L sample per well