Learn More
New England Biolabs, Inc. PhiX174 DNA-Hae III Digest – 0.25 ml
SDP
Catalog No. 50811762
Encompass
The HaeIII digest of X174 yields 11 fragments suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-811-762 Supplier New England Biolabs, Inc. Supplier No. N3026L
The HaeIII digest of X174 yields 11 fragments suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.