Learn More
Sigma Aldrich Fine Chemicals Biosciences phiX174 DNA Marker Hae III1MG
Catalog No. 502905532
Encompass
Sigmas Phi X174 DNA Marker Hae III Digest contains 11 fragments from 72 - 1 353 bp. five ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.
Catalog No. 50-290-5532 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D0672.1MG
Sigmas Phi X174 DNA Marker Hae III Digest contains 11 fragments from 72 - 1 353 bp. five ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.