Learn More
New England Biolabs, Inc. PhiX174 RF I DNA – 30 µg
SDP
Catalog No. 50811757
Encompass
This is the double-stranded, covalently closed, circular form of X174 RF I DNA (supercoiled). The molecular weight of X174 RF I is 3.50 x 10^6 daltons and it is 5,386 base pairs in length. The vast majority of the molecules are RF I, and the remainder are RF II, as determined by agarose gel electrophoresis.
Catalog No. 50-811-757 Supplier New England Biolabs, Inc. Supplier No. N3021S
This is the double-stranded, covalently closed, circular form of X174 RF I DNA (supercoiled). The molecular weight of X174 RF I is 3.50 x 10^6 daltons and it is 5,386 base pairs in length. The vast majority of the molecules are RF I, and the remainder are RF II, as determined by agarose gel electrophoresis.