Learn More
New England Biolabs, Inc. PhiX174 RF II DNA – 150 µg
SDP
Catalog No. 50811758
Encompass
This commonly used cloning vector is the double-stranded nicked circular form of ΦX174 - relaxed form. Its molecular weight is 3.5 x10^6 Daltons and it is 5,386 base pairs in length. Greater then 90% of the molecules are RF II, the remainder are RF I (supercoiled) and linear ΦX174 DNA.
Catalog No. 50-811-758 Supplier New England Biolabs, Inc. Supplier No. N3022L
This commonly used cloning vector is the double-stranded nicked circular form of ΦX174 - relaxed form. Its molecular weight is 3.5 x10^6 Daltons and it is 5,386 base pairs in length. Greater then 90% of the molecules are RF II, the remainder are RF I (supercoiled) and linear ΦX174 DNA.