Learn More
New England Biolabs, Inc. PhiX174 Virion DNA – 250 µg
SDP
Catalog No. 50811760
Encompass
ΦX174 Virion DNA is the single-stranded viral DNA isolated from bacteriophage ΦX174 am3 cs70. It has a molecular weight of 1.70 x 10^6 daltons and is 5386 bases in length. Greater than 85% of the molecules are circular.
Catalog No. 50-811-760 Supplier New England Biolabs, Inc. Supplier No. N3023L
ΦX174 Virion DNA is the single-stranded viral DNA isolated from bacteriophage ΦX174 am3 cs70. It has a molecular weight of 1.70 x 10^6 daltons and is 5386 bases in length. Greater than 85% of the molecules are circular.