Learn More
Aviva Systems Biology Corp Phkg1 Antibody - N-terminal region (ARP56680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Phkg1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Phkg, PHKIN01, Phkg1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-192