Learn More
Aviva Systems Biology Corp PHKG2 Antibody - N-terminal region (ARP33072_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33072_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHKG2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PHKG2. Alternate names: GSD9CApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54873