Learn More
Aviva Systems Biology Corp PHLDA2 Antibody - N-terminal region (ARP58238_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58238_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHLDA2. It was validated on Western Blot. . Alternate names: BRW1C, BWR1C, HLDA2, IPL, TSSC3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rat
Catalog No. 50-286-455