Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Phlebotomy Draw Station

Catalog No. 22505339
Change view
Click to view available options
Dimensions (L x W x H):
15 x 13 x 3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22-505-339 15 x 13 x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-339 Supplier Marketlab Supplier No. 7250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to support efficient phlebotomy workflows.

  • Keep all your supplies close at hand
  • Organize small supplies with several compartments
  • Includes 2 tube racks for 13 mm and 17 mm tubes

Specifications

Dimensions (L x W x H) 15 x 13 x 3 in.
Material Acrylic
Product Type Phlebotomy Tray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.