Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Phlebotomy Super Station on Steel Stand
SDP

Catalog No. 22000769
Change view
Click to view available options
Holds:
Vials, Vacutainer, Tape, Tourniquet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-000-769 Vials, Vacutainer, Tape, Tourniquet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-769 Supplier Poltex™ Supplier No. PHLEBSTATMONST3STL
Add to Cart
Add to Cart

Phlebotomy super organizer on steel stand.

  • Holds any item you might need for lab draw procedure
  • Comes with powder coated steel stand
  • Super durable PETG construction

Specifications

Product Type Organizer
Includes 3 Tape, 3 Tourniquet Rods
Width (Metric) 368 mm
For Use With (Application) Phlebotomy Supplies
Color Clear
Autoclavable Not Autoclavable
Holds Vials, Vacutainer, Tape, Tourniquet
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.