Learn More
Medicus Health Phlebotomy Tray Tube Rack
SDP
Catalog No. NC3790343
Encompass
• Phlebotomy Tray Tube Rack fits inside Phlebotomy Tray with 8 Bins Cups and holds 28 blood draw tubes
• Dimensions: 4.5" L x 3" W x 3" H
• Epoxy coated wire tube rack holds 28 blood draw tubes
• For tubes up to 13mm in diameter
Catalog No. NC3790343 Supplier Medicus Health Supplier No. 5524M2
• Phlebotomy Tray Tube Rack fits inside Phlebotomy Tray with 8 Bins Cups and holds 28 blood draw tubes
• Dimensions: 4.5" L x 3" W x 3" H
• Epoxy coated wire tube rack holds 28 blood draw tubes
• For tubes up to 13mm in diameter