Learn More
Invivogen Phleomycin, 100 mg
SDP
Catalog No. NC9198593
Encompass
Phleomycin is a glycopeptide antibiotic of the bleomycin family, isolated from a mutant strain of Streptomyces verticillus. It binds and intercalates DNA thus destroying the integrity of the double helix. Phleomycin is active against most bacteria, filamentous fungi, yeast, plant and animal cells.
Catalog No. NC9198593 Supplier Invivogen Supplier No. ANTPH1
Phleomycin is a glycopeptide antibiotic of the bleomycin family, isolated from a mutant strain of Streptomyces verticillus. It binds and intercalates DNA thus destroying the integrity of the double helix. Phleomycin is active against most bacteria, filamentous fungi, yeast, plant and animal cells.