Learn More
Alkali Scientific Phleomycin 20Mg/mL Solution, 5 Milliliters
SDP
Supplier:  Alkali Scientific PHL001CP
Encompass_Preferred
Phleomycin 20mg/mL Solution, 5mL is a ready-to-use solution of an antibiotic agent that is used in cancer research, particularly in studies involving DNA damage and chemotherapy. Phleomycin works by inducing DNA strand breaks, making it effective in the treatment of various cancers, particularly those involving solid tumors. This ready-to-use format simplifies the use of Phleomycin in laboratory experiments, reducing preparation time and ensuring precise dosages. The 5mL vial is ideal for small-scale studies or experiments, and the 20mg/mL concentration ensures that the solution is potent and effective for most laboratory applications involving DNA and cell studies.
Catalog No. 50-006-61376