Learn More
Research Products International Corp Phleomycin 20mg/ml Solution, 5 ML
SDP
Catalog No. 501977175
Encompass_Preferred
Phleomycin is an antibiotic and antitumor agent commonly used in molecular biology and life science research. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms. 
Phleomycin 20mg/ml Solution is a formulation often employed in molecular biology experiments to induce controlled DNA damage in cells. Phleomycin accomplishes this by causing single- and double-strand breaks in DNA, making it a valuable tool for investigating cellular responses to DNA damage and the mechanisms involved in DNA repair. 
Researchers may use Phleomycin 20mg/ml Solution to study the impact of DNA damage on cellular processes, gene expression, and other aspects of molecular biology.
Catalog No. 50-197-7175 Supplier Research Products International Corp Supplier No. P202205.0
Phleomycin is an antibiotic and antitumor agent commonly used in molecular biology and life science research. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms. 
Phleomycin 20mg/ml Solution is a formulation often employed in molecular biology experiments to induce controlled DNA damage in cells. Phleomycin accomplishes this by causing single- and double-strand breaks in DNA, making it a valuable tool for investigating cellular responses to DNA damage and the mechanisms involved in DNA repair. 
Researchers may use Phleomycin 20mg/ml Solution to study the impact of DNA damage on cellular processes, gene expression, and other aspects of molecular biology.