Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phloracetophenone | 480-66-0 | 168.15 | 1 G
SDP

Catalog No. 5000447885
Encompass_Preferred

Phloracetophenone is a chemical compound often used as a laboratory chemical and in the manufacture of various substances. Web snippets indicate it acts as a Cytochrome P450 Activator and exhibits antibacterial properties. This product is a solid with a molecular weight of 168.15.

  • Appearance: Solid
  • Melting point: 219-221 °C
  • Boiling point: 257.07 °C at 760 mmHg
  • Identified for laboratory research

Catalog No. 50-004-47885 Supplier Medchemexpress LLC Supplier No. HYW0082261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phloracetophenone is a chemical compound often used as a laboratory chemical and in the manufacture of various substances. Web snippets indicate it acts as a Cytochrome P450 Activator and exhibits antibacterial properties. This product is a solid with a molecular weight of 168.15.

  • Appearance: Solid
  • Melting point: 219-221 °C
  • Boiling point: 257.07 °C at 760 mmHg
  • Identified for laboratory research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.