Learn More
Apexbio Technology LLC Phloretin 60-82-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063519
Encompass_Preferred
Phloretin (CAS 60-82-2) is a naturally occurring dihydrochalcone phenolic compound derived primarily from apple leaves and Manchurian apricot It functions as an inhibitor of sodium-glucose linked transporters (SGLT1 and SGLT2) proteins involved in glucose uptake and reabsorption in intestinal and renal tissues Phloretin inhibits glucose uptake via these transporters exhibiting an IC50 of approximately 49 12 M In cell-based studies it enhances lipolysis increases AMP-activated protein kinase (AMPK) phosphorylation and suppresses adipogenic transcription factors and MAPK signaling Animal research demonstrates that oral administration of Phloretin reduces blood glucose and improves lipid profiles in diabetic mouse models highlighting its utility in metabolic research
Catalog No. 50-000-63519 Supplier Apexbio Technology LLC Supplier No. A37235.1
Phloretin (CAS 60-82-2) is a naturally occurring dihydrochalcone phenolic compound derived primarily from apple leaves and Manchurian apricot It functions as an inhibitor of sodium-glucose linked transporters (SGLT1 and SGLT2) proteins involved in glucose uptake and reabsorption in intestinal and renal tissues Phloretin inhibits glucose uptake via these transporters exhibiting an IC50 of approximately 49 12 M In cell-based studies it enhances lipolysis increases AMP-activated protein kinase (AMPK) phosphorylation and suppresses adipogenic transcription factors and MAPK signaling Animal research demonstrates that oral administration of Phloretin reduces blood glucose and improves lipid profiles in diabetic mouse models highlighting its utility in metabolic research