Learn More
Enzo Life Sciences Phloretin (200mg). CAS: 60-82-2
SDP
Catalog No. 502010575
Encompass_Preferred
Flavonoid. Protein kinase C (PKC) inhibitor. PGF2a receptor antagonist. In astrocytes, these receptors are linked to phospholipase C, thus affecting phosphoinositide hydrolysis and intracellular Ca2+ levels. Inhibits myo-inositol uptake and 5’-iodothyronine deiodinase. Prevents TNF-α stimulated upregulation of VCAM-1, ICAM-1 and E-selectin. Enhances adiponectin expression. Purity: ≥98% (TLC). Solubility: Soluble in acetone, DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0575 Supplier Enzo Life Sciences Supplier No. BMLEI1540200
Flavonoid. Protein kinase C (PKC) inhibitor. PGF2a receptor antagonist. In astrocytes, these receptors are linked to phospholipase C, thus affecting phosphoinositide hydrolysis and intracellular Ca2+ levels. Inhibits myo-inositol uptake and 5’-iodothyronine deiodinase. Prevents TNF-α stimulated upregulation of VCAM-1, ICAM-1 and E-selectin. Enhances adiponectin expression. Purity: ≥98% (TLC). Solubility: Soluble in acetone, DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.