Learn More
Medchemexpress LLC Phloretin | 60-82-2 | 99.92% | 274.27 | 50 G
SDP
Catalog No. 5000427223
Encompass_Preferred
Phloretin is a flavonoid extracted from *Malus pumila Mill.* that possesses anti-inflammatory activities. It is also a specific, competitive, and orally active inhibitor of sodium/glucose cotransporters in the intestine (SGLT1) and kidney (SGLT2). Phloretin inhibits Yeast-made GLUT1 and Human erythrocyte GLUT1 with IC50 values of 49 μM and 61 μM, respectively. It has potential for the treatment of rheumatoid arthritis (RA) and allergic airway inflammation.
  • Possesses anti-inflammatory activities
  • Inhibits SGLT1 and SGLT2
  • Inhibits Yeast-made GLUT1 and Human erythrocyte GLUT1
  • Potential for treating rheumatoid arthritis and allergic airway inflammation
Catalog No. 50-004-27223 Supplier Medchemexpress LLC Supplier No. HYN014250G
Phloretin is a flavonoid extracted from *Malus pumila Mill.* that possesses anti-inflammatory activities. It is also a specific, competitive, and orally active inhibitor of sodium/glucose cotransporters in the intestine (SGLT1) and kidney (SGLT2). Phloretin inhibits Yeast-made GLUT1 and Human erythrocyte GLUT1 with IC50 values of 49 μM and 61 μM, respectively. It has potential for the treatment of rheumatoid arthritis (RA) and allergic airway inflammation.
  • Possesses anti-inflammatory activities
  • Inhibits SGLT1 and SGLT2
  • Inhibits Yeast-made GLUT1 and Human erythrocyte GLUT1
  • Potential for treating rheumatoid arthritis and allergic airway inflammation