Learn More
Phloridzin dihydrate, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 7061-54-3 |
| Physical Form | Neat |
| Linear Formula | C21H24O10 · 2H2 O |
| MDL Number | MFCD00149438 |
| Melting Point | 113°C to 114°C |
| Synonym | 1-[2-(beta-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone; Phloretin 2 ′-beta-D-glucopyranoside; Phloretin 2 ′-beta-D-glucoside; Phlorizin dihydrate |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 472.44 |
| Percent Purity | ≥98.5% (HPLC) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.