Learn More
Selleck Chemical LLC Phlorizin S2343-1g
SDP
Catalog No. 503112794
Encompass_Preferred
Phlorizin (Phloridzin) is a dihydrochalcone found in the bark of pear (Pyrus communis) apple cherry and other fruit trees Phlorizin is a non-selective SGLT inhibitor with Ki of 300 nM and 39 nM for hSGLT1 and hSGLT2 respectively Phlorizin is also a Na /K -ATPase inhibitor
Catalog No. 50-311-2794 Supplier Selleck Chemical LLC Supplier No. S23431G
Phlorizin (Phloridzin) is a dihydrochalcone found in the bark of pear (Pyrus communis) apple cherry and other fruit trees Phlorizin is a non-selective SGLT inhibitor with Ki of 300 nM and 39 nM for hSGLT1 and hSGLT2 respectively Phlorizin is also a Na /K -ATPase inhibitor