Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phlorizin | 60-81-1 | MFCD00006652 | ≥98% | 436.43 | 1 G
SDP

Catalog No. 5000365380
Encompass_Preferred

Phlorizin is a biochemical reagent that acts as a non-selective SGLT inhibitor (hSGLT1 Ki: 300 nM, hSGLT2 Ki: 39 nM) and a Na+/K+-ATPase inhibitor. It is suitable for use as a biological material or organic compound in life science research.

  • Non-selective SGLT inhibitor
  • Na+/K+-ATPase inhibitor
  • Classification: Flavonoids, Phenols, Polyphenols, Dihydrochalcones
  • Source: Malus pumila Mill. (Rosaceae), Plants
  • Targets: SGLT, Na+/K+ ATPase
  • Research areas: Metabolic disease

Catalog No. 50-003-65380 Supplier Medchemexpress LLC Supplier No. 1GHYN0143
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phlorizin is a biochemical reagent that acts as a non-selective SGLT inhibitor (hSGLT1 Ki: 300 nM, hSGLT2 Ki: 39 nM) and a Na+/K+-ATPase inhibitor. It is suitable for use as a biological material or organic compound in life science research.

  • Non-selective SGLT inhibitor
  • Na+/K+-ATPase inhibitor
  • Classification: Flavonoids, Phenols, Polyphenols, Dihydrochalcones
  • Source: Malus pumila Mill. (Rosaceae), Plants
  • Targets: SGLT, Na+/K+ ATPase
  • Research areas: Metabolic disease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.