Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phlorizin Hydrate 97.0+%, TCI America™
SDP

Catalog No. P02481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02481G 1 g
P02485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02481G Supplier TCI America Supplier No. P02481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-81-1
Molecular Formula C21H24O10
Molecular Weight (g/mol) 436.413
MDL Number MFCD00006591
InChI Key IOUVKUPGCMBWBT-QNDFHXLGSA-N
Synonym phlorizin, phloridzin, phlorhizin, phlorizoside, floridzin, phlorrhizin, phloretin 2'-glucoside, phloridzosid, phlorrhizen, phlorizine
PubChem CID 6072
ChEBI CHEBI:8113
IUPAC Name 1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
SMILES C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Specifications

Quantity 1 g
Formula Weight 436.41
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phlorizin Hydrate
RTECSNumber UC2080000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.