Learn More
Cayman Chemical Phloroglucinol, Purity: ≥98%, CAS: 108-73-6, 25 g
SDP
Catalog No. 502987215
Encompass_Preferred
A phenol with diverse biological activities; protects V79-4 Chinese hamster lung fibroblast cells from oxidative stress and inhibits lipid peroxidation by scavenging ROS; induces apoptosis in HT-29 human colon cancer cells; inhibits metastasis of BT549 and MDA-MB-231 human breast cancer cells; protects primary neurons from β-amyloid-induced dendritic spine loss; shortens the latency to find the platform in a Morris water maze test in an AD mouse model; used to stain histological plant sections and in the synthesis of numerous natural products; slows the frequency and decreases the amplitude of contraction in isolated rabbit and rat intestine at a concentration of 100 μM and 1 μM, respectively
Catalog No. 50-298-7215 Supplier Cayman Chemical Supplier No. 2322325 G
A phenol with diverse biological activities; protects V79-4 Chinese hamster lung fibroblast cells from oxidative stress and inhibits lipid peroxidation by scavenging ROS; induces apoptosis in HT-29 human colon cancer cells; inhibits metastasis of BT549 and MDA-MB-231 human breast cancer cells; protects primary neurons from β-amyloid-induced dendritic spine loss; shortens the latency to find the platform in a Morris water maze test in an AD mouse model; used to stain histological plant sections and in the synthesis of numerous natural products; slows the frequency and decreases the amplitude of contraction in isolated rabbit and rat intestine at a concentration of 100 μM and 1 μM, respectively