Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol Dihydrate, Reagents

Catalog No. 01337132
Change view
Click to view available options
:
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-132 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-132 Supplier Reagents Holdings Llc Supplier No. C2271300100C2
Only null left
Add to Cart
Add to Cart

Phloroglucinol Dihydrate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym 1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate
IUPAC Name benzene-1,3,5-triol dihydrate
SMILES O.O.OC1=CC(O)=CC(O)=C1

Specifications

Color White or Off-White
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.46
Grade Reagent
Physical Form Solid
Chemical Name or Material Phloroglucinol Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.