Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol TS, 2% (w/v) Alcoholic, Ricca Chemical
SDP

Catalog No. 577532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5775-32 1 L Amber Glass Bottle
5775-16 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5775-32 Supplier Ricca Chemical Company Supplier No. 577532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6099-90-7 , 67-63-0 , 64-17-5
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol dihydrate
SMILES O.O.OC1=CC(O)=CC(O)=C1

Specifications

Color Brown-Yellow to Colorless
CAS Min % 2.39, 86.0, 4.57
CAS Max % 2.49, 90.0, 4.75
Percent Purity 2%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Alcoholic Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Phloroglucinol, Test Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.