Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phloxine B, 85%, pure, high purity, Biological stain

Catalog No. AC189470250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189470250 25 g Glass bottle
AC189470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189470250 Supplier Thermo Scientific Chemicals Supplier No. 189470250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18472-87-2 | C20H2Br4Cl4Na2O5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20H2Br4Cl4Na2O5
MDL Number MFCD00005061
Synonym Acid Red 92, C.I. 45410

Specifications

Absorbance 875 to 1300 (at lambda max.)
Absorption Ratio 1.38 to 2.24
Dye Content 80% min. (on anhydrous substance)
Infrared Spectrum Authentic
Lambda Max 546 to 550nm (in 50:50 METHS:water)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Assay Percent Range 85%
Beilstein 19, IV, 2921
Merck Index 15, 7440
Color Brown to Brown/Red
Quantity 25 g
Physical Form Powder
Formula Weight 829.64
Percent Purity 85%
Grade Pure
Chemical Name or Material Phloxine B
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.