Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloxine B (Certified Biological Stain), Fisher Chemical

Catalog No. P38725 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P387-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. P387-25

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in histology

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20H2Br4 Cl4Na2O5
MDL Number MFCD00070627
Synonym Acid Red 92

Specifications

Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Packaging Glass Bottle
Color Index No. 45410
Color Brown/Red
pH 9.7
Quantity 25 g
Physical Form Powder
Formula Weight 829.66
Grade Certified Biological Stain
Chemical Name or Material Phloxine B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.