Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloxine B, Certified, Sigma-Aldrich

Catalog No. 01259041 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-041 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-041 Supplier MilliporeSigma Supelco Supplier No. P403025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phloxine B is part of the xanthylium class of dyes. The stain is mostly used to determine cell ploidy and temperature sensitivity of Schizosaccharomyces pombe. Solid media is colored a vivid pink.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-87-2
Molecular Weight (g/mol) 829.63 g/mol

Specifications

Color Dark Red
Quantity 25 g
Percent Purity ≥80 %
Grade Certified
Chemical Name or Material Phloxine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.