Learn More
Electron Microscopy Sciences Phloxine B Solution 0.5% 500 ML
SDP
Catalog No. 5031704
Encompass_Preferred
Used in Thomas Method for Malarial Parasites, Thomas (1953). References: Thomas, J.T. Stain Techn., 28:311-312, 1953 and Luna, L.G. (ed.) Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, New York, P.129, c.1968
Catalog No. 50-317-04 Supplier Electron Microscopy Sciences Supplier No. 2615501
Used in Thomas Method for Malarial Parasites, Thomas (1953). References: Thomas, J.T. Stain Techn., 28:311-312, 1953 and Luna, L.G. (ed.) Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, New York, P.129, c.1968