Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloxine B, Spectrum™ Chemical
SDP

Catalog No. 18612306 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-306 25 g Amber Glass Bottle
18-612-304 10 g Amber Glass Bottle
18-612-305 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-612-306 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PH17025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18472-87-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Brown to Red
Quantity 25 g
Molecular Weight (g/mol) 829.66
Percent Purity 85%
Grade Reagent
Chemical Name or Material Phloxine B

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.