Learn More
Sino Biological Recombinant Human PTPRE Protein (GST Tag), Active
Catalog No. 502573620
Encompass_Preferred
Recombinant human PTPRE (75-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3620 Supplier Sino Biological Supplier No. P5221G500
Recombinant human PTPRE (75-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.