Learn More
Medchemexpress LLC Phocaecholic acid (β-phocaecholic acid) | 105369-89-9 | 408.57 | 5 MG
SDP
Catalog No. 5000416456
Encompass_Preferred
Phocaecholic acid (β-Phocaecholic acid) is a bile acid found in duck bile. It can be excreted into bile and partially decarboxylated to nor-chenodeoxycholic acid in rats. This compound is promising for research related to bile acid metabolism and associated diseases. It is for research use only and not sold to patients. The product appears as a white to off-white solid. Its purity is 98.0%.
  • Found in duck bile.
  • Can be excreted into bile and partially decarboxylated to nor-chenodeoxycholic acid in rats.
  • Promising for research related to bile acid metabolism and associated diseases.
  • Appears as a white to off-white solid.
  • Purity is 98.0%.
  • For research use only.
Catalog No. 50-004-16456 Supplier Medchemexpress LLC Supplier No. HYN153175MG
Add to Cart
Add to Cart
Phocaecholic acid (β-Phocaecholic acid) is a bile acid found in duck bile. It can be excreted into bile and partially decarboxylated to nor-chenodeoxycholic acid in rats. This compound is promising for research related to bile acid metabolism and associated diseases. It is for research use only and not sold to patients. The product appears as a white to off-white solid. Its purity is 98.0%.
  • Found in duck bile.
  • Can be excreted into bile and partially decarboxylated to nor-chenodeoxycholic acid in rats.
  • Promising for research related to bile acid metabolism and associated diseases.
  • Appears as a white to off-white solid.
  • Purity is 98.0%.
  • For research use only.