Learn More
HOYA™ Phoenix 1.53
Catalog No. 18700759
Change view
Click to view available options
For Use With (Application):
Moderate to medium myopes and hyperopes
Catalog No. For Use With (Application)
18-700-759 Moderate to medium myopes and hyperopes
1 options
This item is not returnable. View return policy
Catalog No. 18-700-759 Supplier HOYA™ Supplier No. HOYAPHX
Add to Cart
Add to Cart
This item is not returnable. View return policy
HOYA™ Phoenix 1.53 is a Trivex-based optical material that provides mechanical strength and robustness without compromise to optical quality.
  • Ideal lens material for people with an active lifestyle, in particularly children, and for those whom confidence in their lenses is the most important factor.
  • Ideal for the most common prescriptions, moderate to medium myopes (up to -5.00) and moderate to medium hyperopes (up to a +5.00).
  • It's mechanical properties make it particularly good for rimless drill mounts and semi-rimless frames.
  • Phoenix is one of only two materials that are recommended for use in children by the American Optometric Association.
  • Additionally, Phoenix provides 100% UVA and UVB protection.
  • Tough and Durable: Phoenix meets all international standards for high-impact resistance and has superior tensile strength.
  • Light and Comfortable: Phoenix is the lightest lens material available, providing unparalleled wearer comfort.
  • Clear and Attractive: Phoenix provides superior optics and is brilliantly clear.
  • Chemically Resistant: Stands up to assault of hair sprays and other substances that are inadvertently applied to the lens surface.
Specifications
Lens Type Chemical-resistant
Lens Tint Clear
For Use With (Application) Moderate to medium myopes and hyperopes
Lens Material Trivex
UV Protection Yes