Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Diagnostics Fill Solutions for Bayer™ Rapidlab™ Analyzers

Catalog No. 23293175 Shop All Phoenix Diagnostics, Inc Products
Change view
Click to view available options
Type:
pH Fill Solution
Quantity:
4 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
23-293-175 pH Fill Solution 4 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-293-175 Supplier Phoenix Diagnostics Supplier No. 03229
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Bayer Rapidlab analyzers.

Fill solutions for Bayer Rapidlab Analyzers.

Specifications

Type pH Fill Solution
For Use With (Application) Clinical chemistry analysis
For Use With (Equipment) Bayer Rapidlab 200 Series, 348, 634, 800 Series
Quantity 4 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.