Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Diagnostics Olympus™ AU Series ISE Reagents

Catalog No. 22293260 Shop All Phoenix Diagnostics, Inc Products
Change view
Click to view available options
Type:
Buffer
Cleaning Solution
High Serum Standard
Internal Reference Solution
Low Serum Standard
Low/High Urine Solution
Mid Standard
NA+/K+ Check Solution
Reference
Wash Solution
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-293-260 Wash Solution
22-293-251 Buffer
22-293-254 Low Serum Standard
22-293-252 Mid Standard
22-293-255 High Serum Standard
22-293-256 Low/High Urine Solution
22-293-253 Reference
22-293-257 Internal Reference Solution
22-293-258 NA+/K+ Check Solution
22-293-259 Cleaning Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-293-260 Supplier Phoenix Diagnostics Supplier No. 21110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Olympus AU Series analyzers

  • For the quantitative determination of sodium, potassium and chloride concentrations in serum and urine samples
  • Solutions available include buffer, mid-standard, reference, low serum standard, high serum standard, low/high urine standard, internal reference fill, Na/K selectivity check, cleaning and wash

Specifications

Type Wash Solution
For Use With (Equipment) Olympus™ AU series analyzer
Capacity (Metric) 6 x 2000 mL
Reagents Wash solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.