Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pholedrine (hydrochloride) | 877-86-1 | 99.6% | 10 MG
SDP

Catalog No. 5000232449
Encompass_Preferred

Pholedrine hydrochloride is the hydrochloride salt of pholedrine, an indirectly acting sympathomimetic amine used in research on adrenergic receptors and cardiovascular responses. It is supplied as a solid research chemical with recommended storage at 4°C (sealed), and solution storage at -80°C for long-term preservation.

  • Hydrochloride salt form for improved stability.
  • Used to study adrenergic receptor activity and cardiovascular effects.
  • Available in small milligram pack sizes suitable for research, e.g. 10 MG.
  • Molecular formula C10H16ClNO; molecular weight 201.69 g/mol.
  • Recommended storage: 4°C sealed; in solution store at -80°C for long-term.

Catalog No. 50-002-32449 Supplier Medchemexpress LLC Supplier No. HY121609A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pholedrine hydrochloride is the hydrochloride salt of pholedrine, an indirectly acting sympathomimetic amine used in research on adrenergic receptors and cardiovascular responses. It is supplied as a solid research chemical with recommended storage at 4°C (sealed), and solution storage at -80°C for long-term preservation.

  • Hydrochloride salt form for improved stability.
  • Used to study adrenergic receptor activity and cardiovascular effects.
  • Available in small milligram pack sizes suitable for research, e.g. 10 MG.
  • Molecular formula C10H16ClNO; molecular weight 201.69 g/mol.
  • Recommended storage: 4°C sealed; in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.