Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pholedrine (hydrochloride) | 877-86-1 | 99.6% | 100 MG
SDP

Catalog No. 5000230563
Encompass_Preferred

Pholedrine (hydrochloride) is an indirectly acting sympathomimetic amine supplied as a solid hydrochloride salt for laboratory research and substance manufacture. It has a reported molecular formula C10H16ClNO and a molecular weight of 201.69 g/mol.

  • CAS 877-86-1
  • Molecular formula C10H16ClNO
  • Molecular weight 201.69 g/mol
  • Appearance solid
  • Recommended storage 4°C, sealed, away from moisture
  • Purity 99.6% (LCMS)

Catalog No. 50-002-30563 Supplier Medchemexpress LLC Supplier No. HY121609A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pholedrine (hydrochloride) is an indirectly acting sympathomimetic amine supplied as a solid hydrochloride salt for laboratory research and substance manufacture. It has a reported molecular formula C10H16ClNO and a molecular weight of 201.69 g/mol.

  • CAS 877-86-1
  • Molecular formula C10H16ClNO
  • Molecular weight 201.69 g/mol
  • Appearance solid
  • Recommended storage 4°C, sealed, away from moisture
  • Purity 99.6% (LCMS)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.