Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pholedrine (hydrochloride) | 877-86-1 | 99.6% | 25 MG
SDP

Catalog No. 5000230434
Encompass_Preferred

Pholedrine hydrochloride is the hydrochloride salt of pholedrine, an adrenergic cardiovascular agent used in research for its hypertensive and mydriatic effects. Supplied as a solid powder with high purity, it is intended for laboratory research applications.

  • CAS number 877-86-1
  • Chemical formula C10H16ClNO
  • Molecular weight 201.69 g·mol⁻¹
  • Purity 99.6%
  • Appearance solid powder
  • Storage 4°C sealed; in solution -80°C (6 months) or -20°C (1 month)
  • Intended use research reagent for adrenergic and cardiovascular studies

Catalog No. 50-002-30434 Supplier Medchemexpress LLC Supplier No. HY121609A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pholedrine hydrochloride is the hydrochloride salt of pholedrine, an adrenergic cardiovascular agent used in research for its hypertensive and mydriatic effects. Supplied as a solid powder with high purity, it is intended for laboratory research applications.

  • CAS number 877-86-1
  • Chemical formula C10H16ClNO
  • Molecular weight 201.69 g·mol⁻¹
  • Purity 99.6%
  • Appearance solid powder
  • Storage 4°C sealed; in solution -80°C (6 months) or -20°C (1 month)
  • Intended use research reagent for adrenergic and cardiovascular studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.