Learn More
Medchemexpress LLC Pholedrine (hydrochloride) | 877-86-1 | 99.6% | 5 MG
SDP
Catalog No. 5000234805
Encompass_Preferred
Pholedrine (hydrochloride) is an indirectly acting sympathomimetic amine used as a diagnostic mydriatic and for cardiovascular and adrenergic research. It is supplied as the hydrochloride salt suitable for topical ophthalmic diagnostic use and laboratory studies.
  • Used as a diagnostic topical eye drop for Horner's syndrome
  • Acts as an indirectly acting sympathomimetic amine
  • Reported high purity suitable for research applications
  • Hydrochloride salt form for improved handling and formulation
Catalog No. 50-002-34805 Supplier Medchemexpress LLC Supplier No. HY121609A5MG
Pholedrine (hydrochloride) is an indirectly acting sympathomimetic amine used as a diagnostic mydriatic and for cardiovascular and adrenergic research. It is supplied as the hydrochloride salt suitable for topical ophthalmic diagnostic use and laboratory studies.
  • Used as a diagnostic topical eye drop for Horner's syndrome
  • Acts as an indirectly acting sympathomimetic amine
  • Reported high purity suitable for research applications
  • Hydrochloride salt form for improved handling and formulation