Learn More
Medchemexpress LLC Pholedrine (hydrochloride) | 877-86-1 | 99.6% | 50 MG
SDP
Catalog No. 5000231491
Encompass_Preferred
Pholedrine hydrochloride is the hydrochloride salt of pholedrine, a sympathomimetic amine that acts on adrenergic receptors and is used in cardiovascular and receptor signaling research. It is supplied for laboratory research use with accompanying safety documentation and a certificate of analysis.
  • Hydrochloride salt form for improved stability and solubility.
  • High purity: 99.6% (LCMS, per COA).
  • Molecular formula C10H16ClNO; molecular weight 201.69 g/mol.
  • Used in adrenergic receptor and cardiovascular pharmacology studies.
  • Available as a 50 MG research pack.
Catalog No. 50-002-31491 Supplier Medchemexpress LLC Supplier No. HY121609A50MG
Pholedrine hydrochloride is the hydrochloride salt of pholedrine, a sympathomimetic amine that acts on adrenergic receptors and is used in cardiovascular and receptor signaling research. It is supplied for laboratory research use with accompanying safety documentation and a certificate of analysis.
  • Hydrochloride salt form for improved stability and solubility.
  • High purity: 99.6% (LCMS, per COA).
  • Molecular formula C10H16ClNO; molecular weight 201.69 g/mol.
  • Used in adrenergic receptor and cardiovascular pharmacology studies.
  • Available as a 50 MG research pack.