Learn More
AdipoGen Phomopsin A
SDP
Catalog No. 502562502
Encompass_Preferred
Chemical. CAS 64925-80-0. Formula C36H45ClN6O12. MW 789.2. Isolated from Phomopsis leptostromiformis. Macrocyclic heptapeptide mycotoxin. Potent anti-mitotic compound that can cause cell death. Microtubule assembly inhibitor. Binds selectively to dimeric tubulin, inhibiting the formation of the microtubule spindle to block cell division. Binds at a site different from the colchicine binding site and overlapping the vinblastine binding site. Inhibits tubulin-dependent GTP hydrolysis. Binds beta-tubulin from higher organisms but not alpha-tubulin or fungal mycelial tubulin. Causes lupinosis a degenerative disorder in livestock fed infected lupins.
Catalog No. 50-256-2502 Supplier AdipoGen Supplier No. AGCN20515M001
Chemical. CAS 64925-80-0. Formula C36H45ClN6O12. MW 789.2. Isolated from Phomopsis leptostromiformis. Macrocyclic heptapeptide mycotoxin. Potent anti-mitotic compound that can cause cell death. Microtubule assembly inhibitor. Binds selectively to dimeric tubulin, inhibiting the formation of the microtubule spindle to block cell division. Binds at a site different from the colchicine binding site and overlapping the vinblastine binding site. Inhibits tubulin-dependent GTP hydrolysis. Binds beta-tubulin from higher organisms but not alpha-tubulin or fungal mycelial tubulin. Causes lupinosis a degenerative disorder in livestock fed infected lupins.