Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phomopsin A | 64925-80-0 | 1 MG
SDP

Catalog No. 5000418313
Encompass_Preferred

Phomopsin A is a cyclic hexapeptide mycotoxin isolated from the fungus Phomopsis leptostomiformis. It acts as a noncompetitive inhibitor of the binding of radiolabeled vincristine to tubulin.

  • Purity: 99.35%
  • Molecular weight: 789.23
  • Chemical formula: C36H45ClN6O12
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Microorganisms, Phomopsis leptostomiformis

Catalog No. 50-004-18313 Supplier Medchemexpress LLC Supplier No. HYN67931MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phomopsin A is a cyclic hexapeptide mycotoxin isolated from the fungus Phomopsis leptostomiformis. It acts as a noncompetitive inhibitor of the binding of radiolabeled vincristine to tubulin.

  • Purity: 99.35%
  • Molecular weight: 789.23
  • Chemical formula: C36H45ClN6O12
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Microorganisms, Phomopsis leptostomiformis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.