Learn More
AdipoGen Phomoxanthone A
SDP
Supplier:  AdipoGen BVT0453M005
Encompass_Preferred
Chemical. CAS 359844-69-2. Formula C38H38O16. MW 750.7. Isolated from fungus Phomopsis sp M4. Mycotoxin Antibacterial against Gram-positive bacteria. Shows antimalarial and antitubercular activity. Anticancer compound. Cytotoxic against several cancer cell lines pro-apoptotic activity. Active against plant pathogene fungi e.g. P. oryza, Phyt. infestans. Activator of murine T lymphocytes, NK cells and macrophages.
Catalog No. 50-256-1908